COM448643797 Combi-Blocks, Inc.

2-Amino-5-(diethylamino)toluene monoHCl

  • MDL number MFCD00035466
  • Cas Number : 2051-79-8
  • Molecular Weight : 214.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9365

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9365
Availability details for
Quantity
  • Availability
  • Add to Cart