COM448643884 Combi-Blocks, Inc.

Methyl2-hydroxy-1,7-naphthyridine-3-carboxylate

  • MDL number MFCD14156086
  • Cas Number : 1124194-67-7
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9463

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9463
Availability details for
Quantity
  • Availability
  • Add to Cart