COM448643987 Combi-Blocks, Inc.

1-(N-Methylpiperidin-4-yl-methyl)piperazine

  • MDL number MFCD02093539
  • Cas Number : 496808-04-9
  • Molecular Weight : 197.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9579
Availability details for
Quantity
  • Availability
  • Add to Cart