COM448644180 Combi-Blocks, Inc.

O-Benzyl-(d)-serine benzyl ester

  • MDL number MFCD03419260
  • Cas Number : 21948-10-7
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9810

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9810
Availability details for
Quantity
  • Availability
  • Add to Cart