COM448644208 Combi-Blocks, Inc.

Valganciclovir HCl

  • MDL number MFCD08460118
  • Cas Number : 175865-60-8
  • Molecular Weight : 390.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9842

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9842
Availability details for
Quantity
  • Availability
  • Add to Cart