COM448644221 Combi-Blocks, Inc.

3-Benzylpyridine

  • MDL number MFCD00006408
  • Cas Number : 620-95-1
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9863

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9863
Availability details for
Quantity
  • Availability
  • Add to Cart