COM448644337 Combi-Blocks, Inc.

N-(2-Bromophenyl)-n'-(1,2-diphenylethylidene)hydrazine

  • MDL number MFCD09991601
  • Cas Number : 945242-95-5
  • Molecular Weight : 365.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9999

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9999
Availability details for
Quantity
  • Availability
  • Add to Cart