COM448644395 Combi-Blocks, Inc.

5-O-(tert-Butyldimethylsilyl)-2,3-o-isoproylidene-d-ribofuranose

  • MDL number MFCD01075716
  • Cas Number : 68703-51-5
  • Molecular Weight : 304.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0060
Availability details for
Quantity
  • Availability
  • Add to Cart