COM448644650 Combi-Blocks, Inc.

6-Formyl-2,3-dimethoxybenzoic acid

  • MDL number MFCD00039572
  • Cas Number : 519-05-1
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0334

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0334
Availability details for
Quantity
  • Availability
  • Add to Cart