COM448644791 Combi-Blocks, Inc.

4-[(3-Hydroxy-1-azetidinyl)methyl]phenylboronic acid hydrochloride

  • MDL number MFCD26383911
  • Molecular Weight : 243.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0483

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0483
Availability details for
Quantity
  • Availability
  • Add to Cart