COM448644836 Combi-Blocks, Inc.

4-Aminomethyl-1-methyl-2-piperidone

  • MDL number MFCD17015880
  • Cas Number : 1234616-57-9
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0533
Availability details for
Quantity
  • Availability
  • Add to Cart