COM448644965 Combi-Blocks, Inc.

N-Ethyl-2-(1-pyrazolyl)acetamide

  • MDL number MFCD18840072
  • Cas Number : 1205076-85-2
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0674

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0674
Availability details for
Quantity
  • Availability
  • Add to Cart