COM448645013 Combi-Blocks, Inc.

4-Bromo-2-hydroxy-5-methoxybenzaldehyde

  • MDL number MFCD23705605
  • Cas Number : 63272-66-2
  • Molecular Weight : 231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0728

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0728
Availability details for
Quantity
  • Availability
  • Add to Cart