COM448645145 Combi-Blocks, Inc.

4-[(1-Azetidinyl)methyl]phenylboronic acid

  • MDL number MFCD26383903
  • Cas Number : 2246662-21-3
  • Molecular Weight : 191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0874

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0874
Availability details for
Quantity
  • Availability
  • Add to Cart