COM448645226 Combi-Blocks, Inc.

(1R,2R)-Ethyl 2-aminocyclopentanecarboxylate

  • MDL number MFCD20719057
  • Cas Number : 114745-46-9
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0957

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0957
Availability details for
Quantity
  • Availability
  • Add to Cart