COM448645260 Combi-Blocks, Inc.

1,2,3,4,5,6,7,8-Octahydroacridine

  • MDL number MFCD00005032
  • Cas Number : 1658-08-8
  • Molecular Weight : 187.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0992

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-0992
Availability details for
Quantity
  • Availability
  • Add to Cart