COM448645674 Combi-Blocks, Inc.

5-(4-Isopropylphenyl)isoxazole-3-carboxylic acid

  • MDL number MFCD07377124
  • Cas Number : 33282-10-9
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1440

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1440
Availability details for
Quantity
  • Availability
  • Add to Cart