COM448645718 Combi-Blocks, Inc.

Bis(2-naphthalenylmethyl)amine

  • MDL number MFCD01111299
  • Cas Number : 47304-99-4
  • Molecular Weight : 297.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1485
Availability details for
Quantity
  • Availability
  • Add to Cart