COM448645960 Combi-Blocks, Inc.

Ethyl 5-chloropyrazine-2-carboxylate

  • MDL number MFCD18070850
  • Cas Number : 54013-04-6
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1745
Availability details for
Quantity
  • Availability
  • Add to Cart