COM448645971 Combi-Blocks, Inc.

tert-Butyl 3-(2-bromoacetyl)piperidine-1-carboxylate

  • MDL number MFCD18250319
  • Cas Number : 1219813-78-1
  • Molecular Weight : 306.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1756
Availability details for
Quantity
  • Availability
  • Add to Cart