COM448646082 Combi-Blocks, Inc.

Methyl 1-methyl-2-oxopyrrolidine-3-carboxylate

  • MDL number MFCD26045228
  • Cas Number : 114724-98-0
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1876

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1876
Availability details for
Quantity
  • Availability
  • Add to Cart