COM448646144 Combi-Blocks, Inc.

3,4'-Dichloropropiophenone

  • MDL number MFCD00000992
  • Cas Number : 3946-29-0
  • Molecular Weight : 203.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1943

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-1943
Availability details for
Quantity
  • Availability
  • Add to Cart