COM448646615 Combi-Blocks, Inc.

Exo-8-boc-8-azabicyclo[3.2.1]octane-3-carboxylic acid methyl ester

  • MDL number MFCD22415199
  • Cas Number : 1204809-88-0
  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2450

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2450
Availability details for
Quantity
  • Availability
  • Add to Cart