COM448646720 Combi-Blocks, Inc.

2,4,6-Tris(perfluorononyl)-1,3,5-triazine

  • MDL number MFCD00042440
  • Cas Number : 57101-59-4
  • Molecular Weight : 1485.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2558

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2558
Availability details for
Quantity
  • Availability
  • Add to Cart