COM448646751 Combi-Blocks, Inc.

4-Chloro-5-propylpyrimidine

  • MDL number MFCD09971219
  • Cas Number : 25199-00-2
  • Molecular Weight : 156.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2593
Availability details for
Quantity
  • Availability
  • Add to Cart