COM448646913 Combi-Blocks, Inc.

4-Chloro-3-nitrobenzenesulfonic acid, sodium salt

  • MDL number MFCD00007496
  • Cas Number : 17691-19-9
  • Molecular Weight : 259.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2767

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2767
Availability details for
Quantity
  • Availability
  • Add to Cart