COM448646937 Combi-Blocks, Inc.

Methyl 2-acetylamino-3-chloropropionate

  • MDL number MFCD04039377
  • Cas Number : 18635-38-6
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2794

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2794
Availability details for
Quantity
  • Availability
  • Add to Cart