COM448646972 Combi-Blocks, Inc.

Ethyl 5-chloro-3-(trifluoromethyl)-1h-indole-2-carboxylate

  • MDL number MFCD12025897
  • Cas Number : 902772-08-1
  • Molecular Weight : 291.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2829
Availability details for
Quantity
  • Availability
  • Add to Cart