COM448647127 Combi-Blocks, Inc.

5-(Trifluoromethyl)-1h-pyrazole-3-carboxamide

  • MDL number MFCD12030791
  • Cas Number : 1276541-94-6
  • Molecular Weight : 179.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2996
Availability details for
Quantity
  • Availability
  • Add to Cart