COM448647168 Combi-Blocks, Inc.

(1R,6R)-Rel-7-boc-3,7-diazabicyclo[4.2.0]octane

  • MDL number MFCD14581307
  • Cas Number : 1251020-47-9
  • Molecular Weight : 212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3041
Availability details for
Quantity
  • Availability
  • Add to Cart