COM448647348 Combi-Blocks, Inc.

4,5-Dibromo-6,7-dihydro-1h-indeno[5,4-b]furan-8(2h)-one

  • MDL number MFCD09955084
  • Cas Number : 196597-77-0
  • Molecular Weight : 332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3236

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3236
Availability details for
Quantity
  • Availability
  • Add to Cart