COM448647401 Combi-Blocks, Inc.

Ethyl 3-ethoxy-2,2-difluoro-3-hydroxypropionate

  • MDL number MFCD00718642
  • Cas Number : 141546-97-6
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3298
Availability details for
Quantity
  • Availability
  • Add to Cart