COM448647442 Combi-Blocks, Inc.

2-Diphenylamino-ethanol

  • MDL number MFCD01727930
  • Cas Number : 6315-51-1
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3340
Availability details for
Quantity
  • Availability
  • Add to Cart