COM448647477 Combi-Blocks, Inc.

Cyclopropyl-piperidin-3-yl-carbamic acid tert-butyl ester

  • MDL number MFCD21098142
  • Cas Number : 250275-24-2
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3375

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3375
Availability details for
Quantity
  • Availability
  • Add to Cart