COM448647640 Combi-Blocks, Inc.

2-Amino-6-(methoxycarbonyl)benzoic acid hydrochloride

  • MDL number MFCD22200908
  • Cas Number : 311310-97-1
  • Molecular Weight : 231.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3548

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3548
Availability details for
Quantity
  • Availability
  • Add to Cart