COM448647870 Combi-Blocks, Inc.

(1S)-4,5-Dimethoxy-1-[(methylamino)methyl]benzocyclobutane HCl

  • MDL number MFCD13194843
  • Cas Number : 866783-13-3
  • Molecular Weight : 243.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3792

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3792
Availability details for
Quantity
  • Availability
  • Add to Cart