COM448647974 Combi-Blocks, Inc.

5-Fluoro-2-methyl-4-nitropyridine n-oxide

  • MDL number MFCD12828024
  • Cas Number : 113209-88-4
  • Molecular Weight : 172.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3906

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-3906
Availability details for
Quantity
  • Availability
  • Add to Cart