COM448648258 Combi-Blocks, Inc.

1-Methyl-1h-benzimidazol-7-amine

  • MDL number MFCD18807405
  • Cas Number : 21577-25-3
  • Molecular Weight : 147.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4214

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4214
Availability details for
Quantity
  • Availability
  • Add to Cart