COM448648353 Combi-Blocks, Inc.

Moxifloxacin

  • MDL number MFCD04117996
  • Cas Number : 151096-09-2
  • Molecular Weight : 401.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid
Inchi Code 1S/C21H24FN3O4/c1-29-20-17-13(19(26)14(21(27)28)9-25(17)12-4-5-12)7-15(22)18(20)24-8-11-3-2-6-23-16(11)10-24/h7,9,11-12,16,23H,2-6,8,10H2,1H3,(H,27,28)/t11-,16+/m0/s1
Inchi Key FABPRXSRWADJSP-MEDUHNTESA-N

Description

Purity 98%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4316

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4316
Availability details for
Quantity
  • Availability
  • Add to Cart