COM448648394 Combi-Blocks, Inc.

4-(Aminomethyl)-1,3-thiazol-2-amine

  • MDL number MFCD08445680
  • Cas Number : 197893-32-6
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4361

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4361
Availability details for
Quantity
  • Availability
  • Add to Cart