COM448648576 Combi-Blocks, Inc.

4-[(Ethylamino)methyl]pyrocatechol hydrobromide

  • MDL number MFCD26383906
  • Cas Number : 1456821-60-5
  • Molecular Weight : 248.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4558

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4558
Availability details for
Quantity
  • Availability
  • Add to Cart