COM448648964 Combi-Blocks, Inc.

D-Threose

  • MDL number MFCD00043042
  • Cas Number : 95-43-2
  • Molecular Weight : 120.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-4964
Availability details for
Quantity
  • Availability
  • Add to Cart