COM448649569 Combi-Blocks, Inc.

Methyl 4-(hydroxymethyl)bicyclo[2.2.2]octane-1-carboxylate

  • MDL number MFCD20275559
  • Cas Number : 94994-15-7
  • Molecular Weight : 198.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-5629

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-5629
Availability details for
Quantity
  • Availability
  • Add to Cart