COM448649590 Combi-Blocks, Inc.

3-Butynyl p-toluenesulfonate

  • MDL number MFCD00041687
  • Cas Number : 23418-85-1
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-5651

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-5651
Availability details for
Quantity
  • Availability
  • Add to Cart