COM448649601 Combi-Blocks, Inc.

N,N-Dimethylmorpholine-2-carboxamide

  • MDL number MFCD11977241
  • Cas Number : 135072-23-0
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-5662

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-5662
Availability details for
Quantity
  • Availability
  • Add to Cart