COM448649694 Combi-Blocks, Inc.

Mephenesin

  • MDL number MFCD00004718
  • Cas Number : 59-47-2
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-5760

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-5760
Availability details for
Quantity
  • Availability
  • Add to Cart