COM448649970 Combi-Blocks, Inc.

Methyl 1-cbz-3-ethylpiperidine-3-carboxylate

  • MDL number MFCD21641314
  • Cas Number : 1363166-02-2
  • Molecular Weight : 305.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6058

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6058
Availability details for
Quantity
  • Availability
  • Add to Cart