COM448650030 Combi-Blocks, Inc.

(3-Propoxyphenyl)methanamine

  • MDL number MFCD07754795
  • Cas Number : 37806-33-0
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6123
Availability details for
Quantity
  • Availability
  • Add to Cart