COM448650060 Combi-Blocks, Inc.

Piperazin-2-ylacetic acid

  • MDL number MFCD08235224
  • Cas Number : 14566-74-6
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6154

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6154
Availability details for
Quantity
  • Availability
  • Add to Cart