COM448650192 Combi-Blocks, Inc.

2-(5-formylfuran-2-yl)benzonitrile

  • MDL number MFCD02630467
  • Cas Number : 299442-23-2
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6296

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6296
Availability details for
Quantity
  • Availability
  • Add to Cart